CH4017895607 ChemBridge Corporation

3-(2,5-dimethyl-1H-pyrrol-1-yl)-4-methylbenzoic acid

  • MDL number MFCD01874279
  • Cas Number : 313701-78-9
  • Molecular Weight : 229.2744

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5953025

Safety Information

MSDS https://www.hit2lead.com/sds/5953025_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart