CH4017898672 ChemBridge Corporation

2-[(1-acetyl-1H-indol-3-yl)amino]benzoic acid

  • MDL number MFCD01548433
  • Cas Number : 352548-58-4
  • Molecular Weight : 294.3047

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6045051

Safety Information

MSDS https://www.hit2lead.com/sds/6045051_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart