CH4017900294 ChemBridge Corporation

7-(2-oxopropoxy)-2H-chromen-2-one

  • MDL number MFCD00449863
  • Cas Number : 36914-75-7
  • Molecular Weight : 218.2054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6088138

Safety Information

MSDS https://www.hit2lead.com/sds/6088138_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart