CH4017901043 ChemBridge Corporation

5-methoxy-2-phenyl-1-benzofuran-3-carboxylic acid

  • MDL number MFCD00427054
  • Cas Number : 300674-03-7
  • Molecular Weight : 268.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6118696

Safety Information

MSDS https://www.hit2lead.com/sds/6118696_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart