CH4017904560 ChemBridge Corporation

[(4,6-dimethylquinolin-2-yl)thio]acetic acid

  • MDL number MFCD02734700
  • Cas Number : 370848-09-2
  • Molecular Weight : 247.3128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6320947

Safety Information

MSDS https://www.hit2lead.com/sds/6320947_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart