CH4017906646 ChemBridge Corporation

3-(4-methoxyphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD01131646
  • Cas Number : 36640-42-3
  • Molecular Weight : 278.3053

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6432837

Safety Information

MSDS https://www.hit2lead.com/sds/6432837_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart