CH4017906785 ChemBridge Corporation

3-(4-methoxyphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02227818
  • Cas Number : 372107-36-3
  • Molecular Weight : 294.3047

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6435118

Safety Information

MSDS https://www.hit2lead.com/sds/6435118_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart