CH4017910557 ChemBridge Corporation

3-[(1-ethyl-1H-benzimidazol-2-yl)thio]propanoic acid

  • MDL number MFCD01874326
  • Cas Number : 51099-68-4
  • Molecular Weight : 250.3168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6706370

Safety Information

MSDS https://www.hit2lead.com/sds/6706370_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart