CH4017936296 ChemBridge Corporation

3-(3,4-dimethoxyphenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD01924091
  • Cas Number : 350997-69-2
  • Molecular Weight : 232.2353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000256

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3000256
Availability details for
Quantity
  • Availability
  • Add to Cart