CH4017936297 ChemBridge Corporation

3-ethoxy-4-isopropoxybenzaldehyde

  • MDL number MFCD01921990
  • Cas Number : 284044-35-5
  • Molecular Weight : 208.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000274

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3000274
Availability details for
Quantity
  • Availability
  • Add to Cart