CH4017936301 ChemBridge Corporation

1-(4-sec-butylphenyl)propan-1-one

  • MDL number MFCD01993685
  • Cas Number : 96187-76-7
  • Molecular Weight : 190.2814

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000312

Safety Information

MSDS https://www.hit2lead.com/sds/3000312_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart