CH4017936310 ChemBridge Corporation

propyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD00449584
  • Cas Number : 302561-09-7
  • Molecular Weight : 239.3339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000503

Safety Information

MSDS https://www.hit2lead.com/sds/3000503_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart