CH4017936314 ChemBridge Corporation

2-amino-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxamide

  • MDL number MFCD01114968
  • Cas Number : 40106-15-8
  • Molecular Weight : 224.3225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000706

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3000706
Availability details for
Quantity
  • Availability
  • Add to Cart