CH4017936319 ChemBridge Corporation

methyl 2-amino-5-phenylthiophene-3-carboxylate

  • MDL number MFCD02090936
  • Cas Number : 61325-02-8
  • Molecular Weight : 233.2862

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3000752

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3000752
Availability details for
Quantity
  • Availability
  • Add to Cart