CH4017936357 ChemBridge Corporation

2-(4-chloro-3,5-dimethylphenoxy)acetohydrazide

  • MDL number MFCD00705188
  • Cas Number : 156867-62-8
  • Molecular Weight : 228.6754

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001435

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3001435
Availability details for
Quantity
  • Availability
  • Add to Cart