CH4017936368 ChemBridge Corporation

4-(allyloxy)-3-methoxybenzaldehyde

  • MDL number MFCD01157220
  • Cas Number : 22280-95-1
  • Molecular Weight : 192.2112

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001530

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3001530
Availability details for
Quantity
  • Availability
  • Add to Cart