CH4017936374 ChemBridge Corporation

4-[(3-chlorobenzyl)oxy]-3-methoxybenzaldehyde

  • MDL number MFCD02255723
  • Cas Number : 588678-16-4
  • Molecular Weight : 276.7149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3003560

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3003560
Availability details for
Quantity
  • Availability
  • Add to Cart