CH4017936382 ChemBridge Corporation

2-bromo-4-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD01119128
  • Cas Number : 60632-40-8
  • Molecular Weight : 231.0434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3004167

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3004167
Availability details for
Quantity
  • Availability
  • Add to Cart