CH4017936404 ChemBridge Corporation

4-(4-ethylphenyl)-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD02730016
  • Cas Number : 438218-98-5
  • Molecular Weight : 218.318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3007461

Safety Information

MSDS https://www.hit2lead.com/sds/3007461_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart