CH4017936443 ChemBridge Corporation

3-methoxy-2-[(3-methylbenzyl)oxy]benzaldehyde

  • MDL number MFCD03422409
  • Cas Number : 588713-63-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014114

Safety Information

MSDS https://www.hit2lead.com/sds/3014114_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart