CH4017936472 ChemBridge Corporation

3-[(3-chlorobenzyl)oxy]-4-methoxybenzaldehyde

  • MDL number MFCD02815468
  • Cas Number : 447428-98-0
  • Molecular Weight : 276.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014183

Safety Information

MSDS https://www.hit2lead.com/sds/3014183_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart