CH4017936494 ChemBridge Corporation

3-ethoxy-4-[(4-fluorobenzyl)oxy]benzaldehyde

  • MDL number MFCD01590573
  • Cas Number : 352455-50-6
  • Molecular Weight : 274.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014231

Safety Information

MSDS https://www.hit2lead.com/sds/3014231_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart