CH4017936496 ChemBridge Corporation

3-ethoxy-4-[(4-methylbenzyl)oxy]benzaldehyde

  • MDL number MFCD02153009
  • Cas Number : 351066-35-8
  • Molecular Weight : 270.323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3014238

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3014238
Availability details for
Quantity
  • Availability
  • Add to Cart