CH4017936505 ChemBridge Corporation

5-(2-thienyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01043763
  • Cas Number : 4683-00-5
  • Molecular Weight : 183.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3014341

Safety Information

MSDS https://www.hit2lead.com/sds/3014341_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart