CH4017936509 ChemBridge Corporation

2-amino-4-(3,4-dimethylphenyl)thiophene-3-carboxamide

  • MDL number MFCD02609525
  • Cas Number : 519016-86-5
  • Molecular Weight : 246.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3014472

Safety Information

MSDS https://www.hit2lead.com/sds/3014472_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart