CH4017936510 ChemBridge Corporation

2-amino-5-methyl-4-(4-methylphenyl)thiophene-3-carboxamide

  • MDL number MFCD02612111
  • Cas Number : 438194-93-5
  • Molecular Weight : 246.3281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3014476

Safety Information

MSDS https://www.hit2lead.com/sds/3014476_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart