CH4017936547 ChemBridge Corporation

2,2-diphenylacetohydrazide

  • MDL number MFCD00544954
  • Molecular Weight : 226.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3014980

Safety Information

MSDS https://www.hit2lead.com/sds/3014980_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart