CH4017936724 ChemBridge Corporation

4-(4-ethylphenyl)butanoic acid

  • MDL number MFCD01462080
  • Cas Number : 5467-53-8
  • Molecular Weight : 192.2542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002818

Safety Information

MSDS https://www.hit2lead.com/sds/4002818_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart