CH4017936728 ChemBridge Corporation

1-(4-methylpyridin-2-yl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD02665159
  • Cas Number : 383136-14-9
  • Molecular Weight : 186.2099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003265

Safety Information

MSDS https://www.hit2lead.com/sds/4003265_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart