CH4017936780 ChemBridge Corporation

2-(1-benzyl-2-methyl-1H-indol-3-yl)ethanamine

  • MDL number MFCD00426556
  • Cas Number : 18690-56-7
  • Molecular Weight : 264.3648

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5102396

Safety Information

MSDS https://www.hit2lead.com/sds/5102396_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart