CH4017936786 ChemBridge Corporation

2-(4-fluorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD00187337
  • Cas Number : 110768-19-9
  • Molecular Weight : 285.2267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5160677

Safety Information

MSDS https://www.hit2lead.com/sds/5160677_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart