CH4017936788 ChemBridge Corporation

10,10-dichlorotricyclo[7.1.0.0~4,6~]decane-5-carboxylic acid

  • MDL number MFCD00448783
  • Cas Number : 1022462-94-7
  • Molecular Weight : 249.1337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5185929

Safety Information

MSDS https://www.hit2lead.com/sds/5185929_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart