CH4017936789 ChemBridge Corporation

2-(3,4-dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

  • MDL number MFCD00404561
  • Cas Number : 294667-04-2
  • Molecular Weight : 295.2894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5186520

Safety Information

MSDS https://www.hit2lead.com/sds/5186520_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart