CH4017936793 ChemBridge Corporation

5-(3-chloro-4-methylphenyl)-2-furaldehyde

  • MDL number MFCD00579049
  • Cas Number : 57666-53-2
  • Molecular Weight : 220.6517

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5249948

Safety Information

MSDS https://www.hit2lead.com/sds/5249948_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart