CH4017936797 ChemBridge Corporation

4-(3-aminopropyl)-1,2,5-oxadiazole-3-carboxylic acid 5-oxide

  • MDL number MFCD00168548
  • Cas Number : 352647-66-6
  • Molecular Weight : 187.1534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5256294

Safety Information

MSDS https://www.hit2lead.com/sds/5256294_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart