CH4017936799 ChemBridge Corporation

N~3~-benzyl-beta-alaninamide

  • MDL number MFCD00769480
  • Cas Number : 16490-80-5
  • Molecular Weight : 178.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5301711

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/5301711
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart