CH4017936818 ChemBridge Corporation

(1H-indol-3-ylmethyl)(2-methoxybenzyl)amine

  • MDL number MFCD01135668
  • Cas Number : 625409-25-8
  • Molecular Weight : 266.3376

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5538918

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5538918
Availability details for
Quantity
  • Availability
  • Add to Cart