CH4017936821 ChemBridge Corporation

2'-formylbiphenyl-2-carboxylic acid

  • MDL number MFCD04039066
  • Cas Number : 6720-26-9
  • Molecular Weight : 226.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5559904

Safety Information

MSDS https://www.hit2lead.com/sds/5559904_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart