CH4017936834 ChemBridge Corporation

(4-nitro-1H-imidazol-1-yl)acetic acid

  • MDL number MFCD00621266
  • Cas Number : 59566-52-8
  • Molecular Weight : 171.1109

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5667358

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5667358
Availability details for
Quantity
  • Availability
  • Add to Cart