CH4017936838 ChemBridge Corporation

(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD00297279
  • Cas Number : 345637-69-6
  • Molecular Weight : 199.1641

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5676868

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5676868
Availability details for
Quantity
  • Availability
  • Add to Cart