CH4017936839 ChemBridge Corporation

(4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD00297272
  • Cas Number : 345637-67-4
  • Molecular Weight : 264.0335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5678493

Safety Information

MSDS https://www.hit2lead.com/sds/5678493_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart