CH4017936841 ChemBridge Corporation

4-(allylamino)benzoic acid

  • MDL number MFCD02252822
  • Cas Number : 53624-18-3
  • Molecular Weight : 177.1998

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5750073

Safety Information

MSDS https://www.hit2lead.com/sds/5750073_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart