CH4017936845 ChemBridge Corporation

6-methyl-8-nitro-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline-4-carboxylic acid

  • MDL number MFCD01190797
  • Cas Number : 354815-34-2
  • Molecular Weight : 274.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5790744

Safety Information

MSDS https://www.hit2lead.com/sds/5790744_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart