CH4017936868 ChemBridge Corporation

1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD02629483
  • Cas Number : 119673-50-6
  • Molecular Weight : 217.2389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5963053

Safety Information

MSDS https://www.hit2lead.com/sds/5963053_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart