CH4017936871 ChemBridge Corporation

4-[(4-chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD01114905
  • Cas Number : 491831-83-5
  • Molecular Weight : 295.6785

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5992149

Safety Information

MSDS https://www.hit2lead.com/sds/5992149_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart