CH4017936880 ChemBridge Corporation

(2Z)-3-(4-methoxyphenyl)-2-phenylacrylic acid

  • MDL number MFCD00598899
  • Cas Number : 19319-32-5
  • Molecular Weight : 254.2806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6104735

Safety Information

MSDS https://www.hit2lead.com/sds/6104735_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart