CH4017936887 ChemBridge Corporation

3-(1-methyl-2-oxopropoxy)-6H-benzo[c]chromen-6-one

  • MDL number MFCD01876702
  • Cas Number : 314744-71-3
  • Molecular Weight : 282.2907

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6148972

Safety Information

MSDS https://www.hit2lead.com/sds/6148972_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart