CH4017936890 ChemBridge Corporation

6-methyl-7-(1-methyl-2-oxopropoxy)-2,3-dihydrocyclopenta[c]chromen-4(1H)-one

  • MDL number MFCD01845139
  • Cas Number : 314743-74-3
  • Molecular Weight : 286.3224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6156859

Safety Information

MSDS https://www.hit2lead.com/sds/6156859_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart