CH4017936900 ChemBridge Corporation

2-imino-4-oxo-1,3-thiazinane-6-carboxylic acid

  • MDL number MFCD02029591
  • Cas Number : 70596-36-0
  • Molecular Weight : 174.1777

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6368072

Safety Information

MSDS https://www.hit2lead.com/sds/6368072_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart