CH4017936917 ChemBridge Corporation

3-(1-allyl-5-oxo-2-thioxoimidazolidin-4-yl)propanoic acid

  • MDL number MFCD02855356
  • Cas Number : 436855-68-4
  • Molecular Weight : 228.2682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6538728

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6538728
Availability details for
Quantity
  • Availability
  • Add to Cart